Learn More
Fisherbrand™ Cardboard Cryo Boxes with Water-Repellant Standard Coating
Product Code. 10637584 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Green
White
Yellow
Height (Metric):
32 mm
50 mm
75 mm
100 mm
Unit Size:
Pack of 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Height (Metric) Unit Size
10637584 White 100 mm Pack of 10
10573815 White 32 mm Pack of 10
10568815 Green 50 mm Pack of 10
11557623 White 50 mm Pack of 10
10231545 Yellow 50 mm Pack of 10
10739884 Green 75 mm Pack of 10
11822483 White 75 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10637584
  • ONLINE EXCLUSIVE PRICE
    81.55€
    50.97€ / pezzi 10
    Risparmia 30.58€
    37% Sconto
View all promotions
 Restricted Item
In stock
Add to basket
Edge
Add to basket
Combine these boxes with different grid inserts to hold a wide range of tube sizes. Fisherbrand™ Cardboard Cryo Boxes with Water-Repellant Standard Coating are temperature resistant down to -80°C, ideal for ultra-low temperature storage.
  • Provide extraordinary flexibility
  • Can be combined with a wide variety of different grid inserts
Specifications
Color White
Material Cardboard
Type Cryo Box
For Use With (Application) Tubes from dia. 6mm to dia. 28.5mm
Holds 6 mm to 28.5 mm Diameter Tubes
Quantity 10 Pack
Dimensions (L x W x H) 133 x 133 x 100 mm
Height (Metric) 100 mm