Learn More
Carl Zeiss™ 100W Halogen Rectangular Filament Bulb
Codice prodotto. 10138478
Cambia vista
Click to view available options
Tensione:
12 V
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tensione unitSize
10138478 12 V Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10138478 Supplier Carl Zeiss™ Supplier No. 3800591660000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Providing excellent illumination for halogen lamps in the laboratory.
  • 12 volts, 100 watts
  • For use in halogen lamps
Specifications
Tipo Microscope Bulb
Da utilizzare con (apparecchiatura) Standard WL Universal, photomikroskop III, IM 35, ICM 405, Axiom at UEM, ACM stand U in lamp housing 100/250, Axioplan (1986) lamp housing 100 No. 467259-9901/447215
Forma Rectangular
Tensione 12 V
Wattage 100 w