Learn More
Zeiss™ Stereomicroscope Stands and Components
Accessories for Zeiss Microscopes
Marca: Zeiss™ 455104-0000-000
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Height: 260mm
- Includes Stand plate, Plastic contrast plate, 2 stage clip
Specifica
Stand S | |
10.2 in. | |
Stand plate, Plastic contrast plate, 2 stage clip |
Carl Zeiss™ stereomicroscope | |
260 mm |