Learn More
Carteolol Hydrochloride, TRC
Product Code. 17900585
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17900585 5 mg 5mg
17970385 100 mg 100mg
17953515 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17900585 Supplier Toronto Research Chemicals Supplier No. TRCC1844505MG
 Restricted Item
Spedizione stimata: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51781-21-6
Molecular Formula C16 H24 N2 O3 . Cl H
Molecular Weight (g/mol) 328.83
Synonym 5-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-2(1H)-quinolinone Hydrochloride, (+/-)-Carteolol Hydrochloride, dl-Carteolol Hydrochloride;
IUPAC Name 5-[3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-1H-quinolin-2-one;hydrochloride
SMILES Cl.CC(C)(C)NCC(O)COc1cccc2NC(=O)CCc12
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C16H24N2O3.ClH/c1-16(2,3)17-9-11(19)10-21-14-6-4-5-13-12(14)7-8-15(20)18-13;/h4-6,11,17,19H,7-10H2,1-3H3,(H,18,20);1H
Formula Weight 328.1554
Percent Purity >95%
Chemical Name or Material Carteolol Hydrochloride
Grade HPLC