Learn More
Cytiva Cassette Foam Sponges
Product Code. 11750615 Shop All Cytiva Products
Change view
Click to view available options
Thickness (English):
0.11 in.
0.25 in.
Thickness (Metric):
3 mm
6 mm
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (English) Thickness (Metric) Unit Size
11750615 0.25 in. 6 mm Pack of 4
10279394 0.11 in. 3 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11750615 Supplier Cytiva Supplier No. TE25
Spedizione stimata: 02-12-2026
These Cassette Foam Sponges are accessory items for transfer tanks.
  • Cassette foam sponges
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) TE 22 Mini Tank Transfer Unit
Type Cassette Foam Sponge
Material Foam
Quantity 4 Pk.
Thickness (English) 0.25 in.
Thickness (Metric) 6 mm