missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cathepsin B Inhibitor IV

Catalog Number p-7082423
349.35 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
1 mg
10 mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
147859-80-1
C19H31N3O6, C19H31N3O6
CA-074 ME; (L-3-trans-(Propylcarbamyl)oxirane-2-carbonyl)-L-isoleucyl-L-proline methyl ester
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
147859-80-1
C19H31N3O6, C19H31N3O6
CA-074 ME; (L-3-trans-(Propylcarbamyl)oxirane-2-carbonyl)-L-isoleucyl-L-proline methyl ester

A cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.
TRUSTED_SUSTAINABILITY
Quantità 10 mg
Descrizione White
Content And Storage -30°C to -10°C
Da utilizzare con (applicazione) A cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.
Peso formulazione 397.46g/mol
Solubilità Soluble in DMSO
Product Type Signal Transduction Reagent
Forma Powder
InChI Key XGWSRLSPWIEMLQ-UHFFFAOYNA-N
SMILES CCCNC(=O)C1OC1C(=O)NC(C(C)CC)C(=O)N1CCCC1C(=O)OC
IUPAC Name methyl 1-(3-methyl-2-{[3-(propylcarbamoyl)oxiran-2-yl]formamido}pentanoyl)pyrrolidine-2-carboxylate
Molecular Weight (g/mol) 397.47
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'