Learn More
Abcam Cathepsin B Inhibitor Screening Kit (Fluorometric)
Product Code. 32111730 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32111730 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32111730 Supplier Abcam Supplier No. ab185438100Test
 Restricted Item
Spedizione stimata: 25-11-2026
Abcam's Cathepsin B Inhibitor Screening Kit (ab185438) utilizes the ability of Cathepsin B to cleave the synthetic AFC based peptide substrate to release AFC, which can be easily quantified using a fluorometer or fluorescence microplate reader.
Abcam's Cathepsin B Inhibitor Screening Kit (ab185438) utilizes the ability of Cathepsin B to cleave the synthetic AFC based peptide substrate to release AFC, which can be easily quantified using a fluorometer or fluorescence microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests
Regulatory Status RUO