missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cathepsin C Polyclonal Antibody
GREENER_CHOICE

Codice prodotto. 15942905
Cambia vista
Click to view available options
Quantità:
100 μL
Dimensione della confezione:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15942905 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15942905 Fornitore Invitrogen™ N. del fornitore PA576379

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Cathepsin C, known also as dipeptidyl aminopeptidase I (DPPI), is a tetrameric lysosomal cysteine peptidase belonging to the papain family. Cathepsin C is involved in intracellular protein degradation and the processing of protein precursors, where it participates in cell growth, neuraminidase activation, and platelet factor XIII activation. Cathepsin C is largely related to other lysosomal cysteine proteinases, including cathepsin B, H and L. Enzymatically, Cathepsin C is capable of sequentially removing dipeptides from the amino terminus, and it requires halide ions, namely chloride ions, and thiols for complete enzymatic activity. Protein levels of Cathepsin C are detected in a variety of tissues, and it is most highly expressed in spleen, kidney, cytotoxic lymphocytes and myeloid cells, where it localizes to the secretory granule compartment. Cathepsin C is initially synthesized as a proenzyme that is rapidly processed to generate two distinct chains that function together as the mature form of the enzyme.
TRUSTED_SUSTAINABILITY

Specifica

Antigene Cathepsin C
Applicazioni Immunocytochemistry, Western Blot
Classificazione Polyclonal
Concentrazione 1 mg/mL
Coniugato Unconjugated
Formulazione PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CTSC
N. accesso geni P53634, P97821
Alias gene AI047818; CATC; Cathepsin C; cathepsin J; CPPI; CTSC; Dipeptidyl peptidase 1; Dipeptidyl peptidase 1 exclusion domain chain; Dipeptidyl peptidase 1 heavy chain; Dipeptidyl peptidase 1 light chain; Dipeptidyl peptidase I; Dipeptidyl peptidase I exclusion domain chain; Dipeptidyl peptidase I heavy chain; Dipeptidyl peptidase I light chain; Dipeptidyl transferase; dipeptidyl-peptidase; dipeptidylpeptidase 1; dipeptidyl-peptidase 1; dipeptidyl-peptidase I; DPP1; DPPI; DPP-I; HMS; JP; JPD; PALS; PDON1; PLS
Simboli geni CTSC
Specie ospite Rabbit
Immunogeno Recombinant fusion protein of human CTSC.
Metodo di purificazione Antigen affinity chromatography
Quantità 100 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 1075, 13032
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.