Learn More
CAY 10566, TRC
Codice prodotto. 30365343
Cambia vista
Click to view available options
Quantità:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
30365343 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30365343 Fornitore Toronto Research Chemicals N. del fornitore TRCC2271255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 3-[4-(2-Chloro-5-fluorophenoxy)-1-piperidinyl]-6-(5-methyl-1,3,4-oxadiazol-2-yl)pyridazine
CAS 944808-88-2
Formula molecolare C18H17ClFN5O2
Quantità 5 mg
Sinonimo CAY 10566, 3-[4-(2-Chloro-5-fluorophenoxy)-1-piperidinyl]-6-(5-methyl-1,3,4-oxadiazol-2-yl)pyridazine
InChI Formula InChI=1S/C18H17ClFN5O2/c1-11-21-24-18(26-11)15-4-5-17(23-22-15)25-8-6-13(7-9-25)27-16-10-12(20)2-3-14(16)19/h2-5,10,13H,6-9H2,1H3
SMILES Cc1oc(nn1)c2ccc(nn2)N3CCC(CC3)Oc4cc(F)ccc4Cl
IUPAC Name 2-[6-[4-(2-chloro-5-fluorophenoxy)piperidin-1-yl]pyridazin-3-yl]-5-methyl-1,3,4-oxadiazole
Molecular Weight (g/mol) 389.81
Peso formulazione 389.1055