Learn More
CD127 Mouse anti-Human, BB700, Clone: HIL-7R-M21, BD Horizon™
Codice prodotto. 15835019
Cambia vista
Click to view available options
Quantità:
100 Tests
25 Tests
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15835019 100 Tests Pezzo
15845019 25 Tests Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15835019 Fornitore BD Biosciences N. del fornitore 566398
Please to purchase this item. Need a web account? Register with us today!
Mouse Monoclonal Antibody
CD127 Monoclonal antibody specifically detects CD127 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifica
Antigene CD127
Applicazioni Flow Cytometry
Classificazione Monoclonal
Clone HIL-7R-M21
Coniugato Brilliant Blue 700 (BB700)
Specie ospite Mouse
Quantità 100 Tests
Status giuridico RUO
Primario o secondario Primary
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Forma Purified
Isotype IgG1 κ