missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CD1A (Human) Recombinant Protein
Human CD1A full-length ORF ( AAH31645, 1 a.a. - 311 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000909-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
- Sequence: MWNWLKEPLSFHVIWIASFYNHSWKQNLVSGWLSDLQTHTWDSNSSTIVFLWPWSRGNFSNEEWKELETLFRIRTIRSFEGIRRYAHELQFEYPFEIQVTGGCELHSGKVSGSFLQLAYQGSDFVSFQNNSWLPYPVAGNMAKHFCKVLNQNQHENDITHNLLSDTCPRFILGLLDAGKAHLQRQVKPEAWLSHGPSPGPGHLQLVCHVSGFYPKPVWVMWMRGEQEQQGTQRGDILPSADGTWYLRATLEVAAGEAADLSCRVKHSSLEGQDIVLYWEHHSSVGFIILAVIVPLLLLIGLALWFRKRCFC
Specifica
AAH31645 | |
CD1a molecule | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD1A | |
GST |
909 | |
Wheat germ expression system | |
25 μg | |
CD1, FCB6, HTA1, R4, T6 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |