missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems CD33-coupled magnetic beads
For capturing the anti-CD33 antibody from various assay systems
dietro preventivo
Specifica
Contenuto kit | The conjugation was achieved by means of the binding between streptavidin and biotin. Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10-14 mol/L Thus, the binding of streptavidin and bi |
---|---|
Content And Storage | -20°C, Avoid light |
Shipping Condition | RT |
Tipo | Magnetic Affinity Bead |
Specie | Human |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17116471
|
ACROBiosystems
MBC-K008-10MG BEADS |
10 mg |
Questo articolo non è attualmente disponibile o è stato dismesso. La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative. |
||||||
17126471
|
ACROBiosystems
MBC-K008-2MG BEADS |
2 mg |
Questo articolo non è attualmente disponibile o è stato dismesso. La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative. |
||||||
Descrizione
The biotinylated CD33 protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-CD33 antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surface area and unique surface coating, which can help you get the best performance and highly reproducible results. This CD33 coupled magnetic beads will bring great convenience with minimum non-specific binding and developed protocols. This ready-to-use product could greatly save your time and hassle.Specifica
The conjugation was achieved by means of the binding between streptavidin and biotin. Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10-14 mol/L Thus, the binding of streptavidin and bi | |
RT | |
Human |
-20°C, Avoid light | |
Magnetic Affinity Bead |