missing translation for 'onlineSavingsMsg'
Learn More

CD38 Antibody (FS02), HRP, R&D Systems™

Product Code. 18666698 Shop All Bio Techne Products
Change view
Click to view available options
Quantità:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
18666698 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18666698 Supplier Novus Biologicals Supplier No. NBP311435H
 Articolo soggetto a restrizioni
Spedizione stimata: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigene CD38
Applicazioni Flow Cytometry, Immunofluorescence, Immunofluorescence
Classificazione Monoclonal
Clone FS02
Coniugato HRP
Formulazione PBS
Alias gene 2'-phospho-ADP-ribosyl cyclase, 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2'-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10
Specie ospite Mouse
Immunogeno Human CD38 (Uniprot: P28907)
Metodo di purificazione Protein A or G purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Apoptosis, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers
Primario o secondario Primary
ID gene (immissione) 952
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Forma Purified
Isotype IgG1 κ
Linea di prodotti Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.