missing translation for 'onlineSavingsMsg'
Learn More

CD39 Mouse anti-Human, PE, Clone: A1 (also known as A1/CD39), BD Biosciences™

Mouse Monoclonal Antibody

Marca:  BD Biosciences 567157

Dettagli aggiuntivi : Peso : 0.00020kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17227844

  • 143.00€ / 25 tests

Spedizione stimata: 10-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

CD39 Monoclonal antibody specifically detects CD39 in Human samples. It is validated for Flow cytometry
Specifica

Specifica

CD39
Monoclonal
PE
A1; ENTPD1; NTPDase-1; ecto-ATPDase 1; ecto-ATPase 1; ecto-apyrase
PHA-activated Human Peripheral Blood Mononuclear Cells
RUO
Human
Mouse BALB/c IgG1 κ
Flow Cytometry
A1 (also known as A1/CD39)
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed.
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali