missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 (Mantle Cell Lymphoma Marker) Mouse, Clone: CD5/2416, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 921-MSM6-P0
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CD5 (Mantle Cell Lymphoma Marker) | |
Cell Surface | |
CD5/2416 | |
Unconjugated | |
Mouse | |
67kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
921 | |
IgG2b κ |
Western Blot, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human CD5 protein (apprx. aa 269-366) | |
293T, Ramos, or MOLT-4 Cells. Tonsil or Lymph Node | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |