missing translation for 'onlineSavingsMsg'
Learn More

CD90/Thy1 Antibody (1aG4/C5), Alexa Fluor™ 750, Novus Biologicals™

Codice prodotto. 18249013 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18249013 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18249013 Fornitore Novus Biologicals N. del fornitore NBP144877AF750

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Mouse, Human (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).
TRUSTED_SUSTAINABILITY

Specifications

Antigene CD90/Thy1
Applicazioni Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classificazione Monoclonal
Clone 1aG4/C5
Coniugato Alexa Fluor 750
Diluizione Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface)
Formulazione 50 mM sodium borate with 0.05% sodium azide
Alias gene CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Simboli geni THY1
Specie ospite Mouse
Immunogeno thymocytes from C3H/Di mice (alelic form: Thy-1.2 positive)
Quantità 0.1 mL
Primario o secondario Primary
ID gene (immissione) 7070
Test di specificità The antibody 1aG4/C5 recognizes alloantigen Thy-1.2 of mouse Thy-1 (CD90); it does not cross-react with Thy-1.1 alloantigen. Thy-1 is a 18 kDa cell surface GPI linked glycoprotein (Immunoglobulin superfamily); in the mouse it is expressed on all thymocytes and peripheral T lymphocytes.
Target Species Mouse, Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.