Learn More
CD90/Thy1 Antibody (CL1028) [Alexa Fluor™ 647], Novus Biologicals™
Product Code. 30295388 Shop All Bio Techne Products
Change view
Click to view available options
Quantità:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30295388 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30295388 Supplier Novus Biologicals Supplier No. NBP252960AF647
Please to purchase this item. Need a web account? Register with us today!
Mouse Monoclonal Antibody
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigene CD90/Thy1
Applicazioni Western Blot
Classificazione Monoclonal
Clone CL1028
Coniugato Alexa Fluor 647
Formulazione 50mM Sodium Borate
Alias gene CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Specie ospite Mouse
Immunogeno This antibody was developed using a recombinant protein derived from P04216, with the exact immunogen sequence remaining proprietary.
Metodo di purificazione Protein A purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primario o secondario Primary
ID gene (immissione) 7070
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Forma Purified
Isotype IgG2b