missing translation for 'onlineSavingsMsg'
Learn More

CD90/Thy1 Antibody (CL1040) [DyLight 594], Novus Biologicals™

Codice prodotto. 30305686 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30305686 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30305686 Fornitore Novus Biologicals N. del fornitore NBP252961DL594

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifica

Antigene CD90/Thy1
Applicazioni Western Blot
Classificazione Monoclonal
Clone CL1040
Coniugato DyLight 594
Formulazione 50mM Sodium Borate
Alias gene CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Specie ospite Mouse
Immunogeno This antibody was developed using a recombinant protein derived from P04216, with the exact immunogen sequence remaining proprietary.
Metodo di purificazione Protein A purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primario o secondario Primary
ID gene (immissione) 7070
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Forma Purified
Isotype IgG1
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.