missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDK11A Polyclonal Antibody
GREENER_CHOICE

Codice prodotto. 16571102
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16571102 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16571102 Fornitore Invitrogen™ N. del fornitore PA598279

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

CDK11A Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

The PITSLRE beta1 protein, a distantly related member of the Cdk family of protein kinases, induces apoptosis after low levels of ectopic expression. Apoptosis, or programmed cell death, is similarly induced by ectopic expression of an amino terminal deletion mutant retaining the catalytic and carboxyterminal domains of PITSLRE beta1, but not by other mutants lacking Histone H1 kinase activity or by other Cdk family members. The terminology for the ten isoforms of the PITSLRE subfamily of proteins is based on the conserved PSTAIRE box region of Cdc2 p34. Depending on which of the PITSLRE genes produce the protein, the cDNA and protein are designated alpha, beta or gamma (i.e., PITSLRE A gene, alpha; PITSLRE B gene, beta and PITSLRE C gene, gamma). Some of the isoforms such as PITSLRE alpha1 (T cells) and PITSLRE beta1 (B cells and brain), are expressed in specific cell types, while others are expressed ubiquitously.
TRUSTED_SUSTAINABILITY

Specifica

Antigene CDK11A
Applicazioni ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classificazione Polyclonal
Concentrazione 1 mg/mL
Coniugato Unconjugated
Formulazione PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CDK11A
N. accesso geni Q9UQ88
Alias gene CDC2L2; CDC2L3; CDK11 p110; CDK11 p46; CDK11 p58; CDK11A; CDK11-p110; CDK11-p46; CDK11-p58; cell division cycle 2-like 2 (PITSLRE proteins); cell division cycle 2-like protein kinase 2; Cell division protein kinase 11A; cyclin dependent kinase 11A; cyclin-dependent kinase 11A; Galactosyltransferase-associated protein kinase p58/GTA; p58GTA; PITSLRE; PITSLRE B; PITSLRE protein kinase beta; PITSLRE serine/threonine-protein kinase CDC2L2; PITSLREB
Simboli geni CDK11A
Specie ospite Rabbit
Immunogeno Recombinant Human Cyclin-dependent kinase 11A protein (5-216AA).
Metodo di purificazione Protein G
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 728642
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.