missing translation for 'onlineSavingsMsg'
Learn More

CDK8 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA565388

Dettagli aggiuntivi : Peso : 0.01000kg

Codice prodotto. 15908144

  • 516.00€ / 100µL
Spedizione stimata: 30-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: LTEEEPDDKGD KKNQQQQQGN NHTNGTGHPG NQDSSHTQGP PLKKVRVVPP TTTSGGLIMT SDYQRSNPHA AYPNPGPSTS QPQSSMGYSA TS Highest antigen sequence identity to the following orthologs - mouse 96%, rat 33%.

CDK8 Polyclonal antibody specifically detects CDK8 in Human samples. It is validated for Immunocytochemistry, Western Blot.
Specifica

Specifica

CDK8
Polyclonal
Unconjugated
Cdk8
Cdk8; CDK8 protein kinase; cell division protein kinase 8; Cell division protein kinase 8-like protein; cyclin dependent kinase 8; cyclin-dependent kinase 8; K35; mediator complex subunit CDK8; Mediator of RNA polymerase II transcription subunit CDK8; MGC126074; MGC126075; protein kinase K35; RGD1560888; wu:fk95h11
Rabbit
Antigen affinity chromatography
RUO
1024
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P49336
Cdk8
Recombinant Human CDK8
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

For Research Use Only