missing translation for 'onlineSavingsMsg'
Learn More

CEACAM2 Antibody (935610) [PerCP], R&D Systems™

Codice prodotto. 18611894 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
18611894 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18611894 Fornitore Novus Biologicals N. del fornitore FAB9214C
 Articolo soggetto a restrizioni
Spedizione stimata: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rat Monoclonal Antibody

CEACAM2 Monoclonal specifically detects CEACAM2 in Mouse samples. It is validated for CyTOF, Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigene CEACAM2
Applicazioni CyTOF, Flow Cytometry
Classificazione Monoclonal
Clone 935610
Coniugato PerCP
Diluizione Flow Cytometry, CyTOF-ready
Formulazione PBS
Alias gene Bgp;Bgp2;Carcinoembryonic antigen-related cell adhesion molecule 2;Ceacam2-L
Specie ospite Rat
Immunogeno Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-2, Gln35-Asp422, Accession # Q925P2
Metodo di purificazione Protein A or G purified from hybridoma culture supernatant
Quantità 0.1 mL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 26367
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Forma Purified
Isotype IgG1
Linea di prodotti Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.