missing translation for 'onlineSavingsMsg'
Learn More

Cefpirome Sulfate, TRC

Codice prodotto. 30137239
Cambia vista
Click to view available options
Quantità:
100 mg
1 g
2 g
Dimensione della confezione:
100mg
1g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30137239 2 g 2g
30133653 1 g 1g
17921215 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30137239 Fornitore Toronto Research Chemicals N. del fornitore TRCC2435002G

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98753-19-6
Formula molecolare C22 H22 N6 O5 S2 . H2 O4 S
Molecular Weight (g/mol) 612.66
Sinonimo Broact, Cefpirome sulfate, HR 810 sulfate, Keiten, 5H-Cyclopenta[b]pyridinium, 1-[[(6R,7R)-7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-6,7-dihydro-, sulfate (1:1) (9CI, ACI)
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate;sulfuric acid
SMILES CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])C[n+]3cccc4CCCc34)\c5csc(N)n5.OS(=O)(=O)O

Specifications

Materiale o nome chimico Cefpirome sulfate
Note sul grado di purezza HPLC
Quantità 2 g
InChI Formula InChI=1S/C22H22N6O5S2.H2O4S/c1-33-26-15(13-10-35-22(23)24-13)18(29)25-16-19(30)28-17(21(31)32)12(9-34-20(16)28)8-27-7-3-5-11-4-2-6-14(11)27;1-5(2,3)4/h3,5,7,10,16,20H,2,4,6,8-9H2,1H3,(H3-,23,24,25,29,31,32);(H2,1,2,3,4)/b26-15-;/t16-,20-;/m1./s1
Peso formulazione 612.0767
Percent Purity >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.