Learn More
Cell Culture Grade Water Tested to USP/EP Sterile Water, Corning™
BIOPHARMA
Codice prodotto. 15353661
Cambia vista
Click to view available options
Quantità:
200 L
Dimensione della confezione:
200L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
15353661 200 L 200L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product should not be used as an injectable or in the preparation of injectables.
Codice prodotto. 15353661 Fornitore Corning Media Tech N. del fornitore 25065LH
 Articolo soggetto a restrizioni
Spedizione stimata: 25-11-2026
Product should not be used as an injectable or in the preparation of injectables.
High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP/EP Sterile Water are used in preparation of purification and chromatography buffers.
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
TRUSTED_SUSTAINABILITY
Specifica
Quantità 200 L