Learn More
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom Microplate
Color:Black; Sterility:Sterile; Lid:With Lid
Marca: Greiner Bio-One 655086
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Compatible with automated systems
- Alphanumeric well coding
- High clarity of microplates for optimal microscopic examinations
- Stackable
- Individu
Specifica
Standard | |
8 x 12 | |
Black | |
96 | |
Sterile | |
392 μL | |
4800 x g (swinging bucket rotor) | |
8 | |
-20°C to +60°C |
With Lid | |
Not Autoclavable | |
Polystyrene | |
25 to 340 μL | |
Cell Culture | |
Flat Bottom (F-well) | |
32 | |
32/Cs. | |
Chimney |