Learn More
- Home
- Products
- Extraction Systems
- Extraction Thimbles
- Sartorius Cellulose Extraction Thimbles
Sartorius Cellulose Extraction Thimbles
Product Code. 30052924 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric) Inside:
19 mm
22 mm
25 mm
26 mm
28 mm
30 mm
33 mm
35 mm
40 mm
43 mm
Length (Metric):
60 mm
70 mm
80 mm
90 mm
94 mm
100 mm
118 mm
123 mm
130 mm
150 mm
205 mm
Wall Thickness:
1.3 mm
1.5 mm
1.7 mm
1.8 mm
Unit Size:
Pack of 25
26 product options available for selection
| Product Code. | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | Unit Size |
|---|---|---|---|---|
| 30052924 | 25 mm | 100 mm | 1.3 mm | Pack of 25 |
| 30052893 | 19 mm | 90 mm | 1.3 mm | Pack of 25 |
| 30052914 | 22 mm | 80 mm | 1.3 mm | Pack of 25 |
| 30052895 | 22 mm | 100 mm | 1.3 mm | Pack of 25 |
| 30052920 | 25 mm | 60 mm | 1.3 mm | Pack of 25 |
| 30052910 | 25 mm | 70 mm | 1.3 mm | Pack of 25 |
| 30052897 | 25 mm | 80 mm | 1.3 mm | Pack of 25 |
| 30052921 | 26 mm | 60 mm | 1.5 mm | Pack of 25 |
| 30052899 | 28 mm | 60 mm | 1.3 mm | Pack of 25 |
| 30052913 | 28 mm | 80 mm | 1.3 mm | Pack of 25 |
| 30052894 | 28 mm | 100 mm | 1.3 mm | Pack of 25 |
| 30052922 | 30 mm | 80 mm | 1.3 mm | Pack of 25 |
| 30052904 | 30 mm | 100 mm | 1.3 mm | Pack of 25 |
| 30052919 | 33 mm | 60 mm | 1.3 mm | Pack of 25 |
| 30052912 | 33 mm | 80 mm | 1.3 mm | Pack of 25 |
| 30052909 | 33 mm | 90 mm | 1.3 mm | Pack of 25 |
| 30052892 | 33 mm | 94 mm | 1.3 mm | Pack of 25 |
| 30052915 | 33 mm | 100 mm | 1.3 mm | Pack of 25 |
| 30052917 | 33 mm | 118 mm | 1.3 mm | Pack of 25 |
| 30052896 | 33 mm | 130 mm | 1.5 mm | Pack of 25 |
| 30052898 | 33 mm | 205 mm | 1.3 mm | Pack of 25 |
| 30052907 | 35 mm | 150 mm | 1.7 mm | Pack of 25 |
| 30052902 | 40 mm | 100 mm | 1.7 mm | Pack of 25 |
| 30052908 | 40 mm | 123 mm | 1.7 mm | Pack of 25 |
| 30052900 | 40 mm | 150 mm | 1.8 mm | Pack of 25 |
| 30052923 | 43 mm | 123 mm | 1.5 mm | Pack of 25 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
This item is not returnable.
View return policy
Product Code.
30052924
Supplier
Sartorius
Supplier No.
FT1201025100
-
127.00€
115.00€ / pezzi 25
Risparmia 12.00€
9% Sconto
Spedizione stimata: 22-01-2027
Eseguire il login per visualizzare lo stock disponibile
Eseguire il login per visualizzare lo stock disponibile
Add to basket
Add to basket
This item is not returnable.
View return policy
Description
- Composed of high-purity cellulose for uncontaminated results
- Engineered for superior mechanical stability to withstand rigorous use
- Designed for a precise fit across a diverse range of extractors
- Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
Specifications
Specifications
| Diameter (Metric) Inside | 25 mm |
| Material | Cellulose |
| Wall Thickness | 1.3 mm |
| Grade | C300 |
| Length (Metric) | 100 mm |
| Product Type | Extraction Thimbles |