Learn More
Sigma Laborzentrifugen™ Adattatori per cestelli rotondi per centrifuga
Use these adapters for a wide variety of applications. Sigma™ Centrifuge Round Bucket Adapters work with all common laboratory tubes for increased centrifugal flexibility.
Marca: Sigma Laborzentrifugen™ 17377
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- For use with Sigma™ centrifuges
Specifica
Round Portoir | |
4 x 50 mL Culture, Tube no. 15151 | |
bucket no. 13190 for Series 3 Centrifuge | |
2/Pk. |