Learn More
Labconco™ CentriVap™ Complete Concentrator
BIOPHARMA
Codice prodotto. 15172995 Sfoglia Tutto Labconco Corporation Prodotti
Cambia vista
Click to view available options
Tipo:
Acid Resistant with Heat Boost
Acid Resistant without Heat Boost
Heat Boost Included
Heat Boost Not Included
Requisiti elettrici:
230 V 50 Hz 6 A
Dimensione della confezione:
Pezzo
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo Requisiti elettrici Dimensione della confezione
15172995 Heat Boost Included 230 V 50 Hz 6 A Pezzo
15103005 Acid Resistant with Heat Boost 230 V 50 Hz 6 A Pezzo
15162995 Heat Boost Not Included 230 V 50 Hz 6 A Pezzo
15142995 Acid Resistant without Heat Boost 230 V 50 Hz 6 A Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 15172995 Fornitore Labconco™ N. del fornitore 7315032
 Articolo soggetto a restrizioni
Spedizione stimata: 24-03-2027
Comes with everything needed to start processing
Uses a combination of centrifugal force, vacuum and heat to speed evaporation.
Includes:
  • Vacuum Concentrator
  • -50°C Cold Trap
  • Aluminum rotor holds 40 microcentrifuge tubes and 12-13mm tubes
  • Chemical-resistant diaphragm vacuum pump
  • CentriZap Strobe Light allows user to view samples while rotor is spinning and lid is closed
Molecular biology, proteomics, genomics, cell biology, microbiology, biochemistry, drug discovery and analytical chemistry
Specifica
Modello Centrivap
Valori nominali elettrici International
Tipo Heat Boost Included
Requisiti elettrici 230 V 50 Hz 6 A