missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP72 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA568398
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Involved in the recruitment of key centrosomal proteins to the centrosome. Provides centrosomal microtubule-nucleation activity on the gamma-tubulin ring complexes (gamma-TuRCs) and has critical roles in forming a focused bipolar spindle, which is needed for proper tension generation between sister chromatids. Required for localization of KIZ, AKAP9 and gamma-tubulin ring complexes (gamma-TuRCs).Specifica
CEP72 | |
Polyclonal | |
Unconjugated | |
CEP72 | |
2610029E11Rik; 4933440J22Rik; centrosomal protein 72; centrosomal protein 72 kDa; centrosomal protein 72kDa; centrosomal protein of 72 kDa; Cep72; KIAA1519 | |
Rabbit | |
affinity chromatography | |
RUO | |
55722, 74470 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9D3R3, Q9P209 | |
CEP72 | |
Synthetic protein corresponding to residues near the C terminal of human CEP72. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |