Learn More
R&D Systems™ CEPT Cocktail Kit
Product Code. 30000439
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000439 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30000439 Supplier R&D Systems™ Supplier No. 7991/1 KIT
  • ONLINE EXCLUSIVE PRICE
    529.37€
    472.50€ / 1 sets
    Risparmia 56.87€
    10% Sconto
 Restricted Item
In stock
Add to basket
Edge
Add to basket
A Tocris product, part of R&D Systems. Cell culture supplement for improving stem cell survival
The CEPT Cocktail Kit is an easy-to-use kit that consists of Chroman 1 (Cat. No. 7163), Emricasan (Cat. No. 7310), Polyamine Supplement (Cat. No. 7739), trans-ISRIB (Cat. No. 5284) and DMSO (Cat. No. 3176). It contains sufficient material for the preparation of 5 L of cocktail and sterile-filtered DMSO for reconstitution. The CEPT Cocktail is a cell culture supplement for improving the survival of self-renewing pluripotent stem cells and their differentiated progeny in stem cell research. It can be used for routine long-term passaging, cryopreservation, single-cell cloning, and organoid formation.
Please refer to the protocol for further information.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Product Line Tocris
Target Compounds for Stem Cell Proliferation & Renewal