Learn More
R&D Systems™ CGP 71683 hydrochloride
Descrizione
CGP 71683 hydrochloride is an extremely selective, non-peptide NPY Y5 receptor antagonist. Displays > 1000-fold selectivity over Y1, Y2 and Y4 receptors; IC50 values are 1.4, 2765, 7187 and 5637 nM at cloned rat Y5, Y1, Y2 and Y4 receptors respectively. Potently inhibits NPY-induced food intake following i.p. administration in diabetic, free-feeding and fasted rats.
Specifica
Specifica
| Quantità | 10 mg |
| Linea di prodotti | Tocris |
| Product Type | CGP 71683 hydrochloride |
| Purezza | >99% |
| Peso molecolare | 512.07 |
| Inibitori | CGP 71683 hydrochloride |
For Research Use Only