missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CGRP Monoclonal Antibody (4901)
GREENER_CHOICE

Codice prodotto. 11503773
Cambia vista
Click to view available options
Quantità:
50 μL
Dimensione della confezione:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11503773 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11503773 Fornitore Invitrogen™ N. del fornitore MA120209

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

Recommended positive controls: rat alpha-CGRP. MA1-20209 has been successfully used in ELISA, radioimmune assay, neutralizing, and immunocytochemistry procedures. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifica

Antigene CGRP
Applicazioni ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Neutralization, Radioimmune Assays (RIA), Western Blot, Immunocytochemistry
Classificazione Monoclonal
Clone 4901
Concentrazione 2.1 mg/mL
Coniugato Unconjugated
Formulazione PBS with 15mM sodium azide; pH 7.4
Gene Calca
N. accesso geni P01256, P01257, P01258, P06881, P41547
Alias gene alpha-type CGRP; CA; Cal1; CAL6; CALC; Calc1; Calca; calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRP-I; Ct; Ctn; Katacalcin; KC; MGC126648; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; RAT calcitonin; RATCAL6
Simboli geni Calca
Specie ospite Mouse
Immunogeno Rat alpha-CGRP peptide. The epitope recognized by the antibody resides within the C-terminal ten amino acids of rat alpha-CGRP.
Metodo di purificazione Purified
Quantità 50 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 24241, 403946, 796
Target Species Human, Rat, Canine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Forma Liquid
Isotype IgG1
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.