Learn More
Lovibond™ Checkit™ Comparators 2-in-1 Test Kits
Codice prodotto. 10739104
Cambia vista
Click to view available options
Range:
6.5 to 8.4 pH, 0 to 1 mg/L Cl2
6.5 to 8.4 pH, 0 to 1 mg/L Cu
6.5 to 8.4 pH, 0 to 4 mg/L Cl2
6.5 to 8.4 pH, 0 to 5 mg/L Br
6.5 to 8.4 pH, 0.1 to 2 mg/L Cl2
Descrizione:
pH Value
Copper
pH Value
Bromine
pH value
Chlorine
pH value
Chlorine
Dimensione della confezione:
Pezzo
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Range Descrizione unitSize
10739104 6.5 to 8.4 pH, 0 to 4 mg/L Cl2 pH value
Chlorine
Pezzo
10631613 6.5 to 8.4 pH, 0.1 to 2 mg/L Cl2 pH value
Chlorine
Pezzo
10739355 6.5 to 8.4 pH, 0 to 1 mg/L Cu pH Value
Copper
Pezzo
11735458 6.5 to 8.4 pH, 0 to 1 mg/L Cl2 pH value
Chlorine
Pezzo
10193202 6.5 to 8.4 pH, 0 to 1 mg/L Cl2 pH value
Chlorine
Pezzo
10461045 6.5 to 8.4 pH, 0 to 5 mg/L Br pH Value
Bromine
Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Codice prodotto. 10739104 Fornitore Lovibond™ N. del fornitore 147026
 Articolo soggetto a restrizioni
Spedizione stimata: 28-08-2026
Aggiungi al carrello
Aggiungi al carrello
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
  • Easy operation and exact reagent dosing
  • Tablet reagents with a minimm guaranteed shelf life of 5/10 years
  • Rotate continuous gradient color wheel to obtain a color match with the reacted sample
  • Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifica
Descrizione pH value
Chlorine
Range 6.5 to 8.4 pH, 0 to 4 mg/L Cl2
Metodo test Colorimetric
Tipo 2 in 1 Test Kit