missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX ChemiBrite™ EP3 Prostanoid Receptor Stable Cell Line

Codice prodotto. 17345169
Encompass
Cambia vista
Click to view available options
Quantità:
2 Vials
Dimensione della confezione:
2 fiale
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
17345169 2 Vials 2 fiale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17345169 Fornitore DiscoverX N. del fornitore HTS092L

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The ChemiBrite™ EP3 Prostanoid Receptor Stable Cell Line measures PTGER3 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.

The ChemiBrite EP3 Prostanoid Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin E2) based activation of PTGER3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTGER3 alongside exogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of PTGER3 , the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifica

Assay Entry Calcium Flux
Linea cellulare 293H
Da utilizzare con (applicazione) Cell Lines
Primary Agonist Prostaglandin E2
Quantità 2 Vials
Target Entry PTGER3
Numero di accesso NM_198716
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.