Learn More
Thermo Scientific™ Chest Freezer Racks
Product Code. 11796479 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
Boxes (2 in.)
Boxes (3 in.)
General Freezer Items
Plates
For Use With (Equipment):
12.7 cu. ft. Freezers
12.7, 17, 20 cu. ft. Freezers
12.7, 20 cu. ft. Freezers
13, 17, 20 cu. ft. Freezers
20 cu. ft. Freezers
3 cu. ft. Chest Freezers
3 cu. ft. Freezers
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds For Use With (Equipment) Unit Size
11796479 General Freezer Items 12.7, 20 cu. ft. Freezers Each
13504940 General Freezer Items 20 cu. ft. Freezers Each
13584930 General Freezer Items 12.7 cu. ft. Freezers Each
12623115 Boxes (2 in.) 3 cu. ft. Chest Freezers Each
12693665 Boxes (3 in.) 3 cu. ft. Freezers Each
11209769 Boxes (2 in.) 13, 17, 20 cu. ft. Freezers Each
12633115 Boxes (3 in.) 13, 17, 20 cu. ft. Freezers Each
11800553 Plates 12.7, 17, 20 cu. ft. Freezers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11796479 Supplier Thermo Scientific™ Supplier No. 114048
  • ONLINE EXCLUSIVE PRICE
    375.22€
    343.00€ / pezzi 1
    Risparmia 32.22€
    8% Sconto
 Restricted Item
Spedizione stimata: 25-05-2027
Use Thermo Scientific™ Chest Freezer Racks with Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Silver
Holds General Freezer Items
Material Stainless Steel
Positions Not Applicable
Shape Rectangular
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) 12.7, 20 cu. ft. Freezers
Freezer Compatibility (Capacity) 12.7 and 20 cu. ft.
Width (English) 11.8 in.
Height (English) 12.3 in.
Depth (English) 16.5 in.
Depth (Metric) 42 cm
Product Type Wire Basket (Rollers)
Shelves None
Height (Metric) 31.2 cm
Width (Metric) 30 cm