Learn More
Chlorambucil, Mikromol™
Description
Chlorambucil
Specifications
Specifications
| Chemical Name or Material | Chlorambucil |
| CAS | 305-03-3 |
| Analyte or Component Names | Chlorambucil |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H19 Cl2 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzenebutanoic acid, 4-[bis(2-chloroethyl)amino]-, Butyric acid, 4-[p-[bis(2-chloroethyl)amino]phenyl]- (8CI), 4-[Bis(2-chloroethyl)amino]benzenebutanoic acid, 4-[p-[Bis(2-chloroethyl)amino]phenyl]butyric acid, Ambochlorin, Amboclorin, CB 1348, Chlorambucil, Chloraminophene, Chlorbutin, Chlorobutine, Ecloril, Leukeran, Leukeran Tablets, Linfolizin, Linfolysin, Lympholysin, NCI 3088, NSC 3088, gamma-[p-Bis(2-chloroethyl)aminophenyl]butyric acid, gamma-[p-Di(2-chloroethyl)aminophenyl]butyric acid |
| InChI Formula | InChI=1S/C14H19Cl2NO2/c15-8-10-17(11-9-16)13-6-4-12(5-7-13)2-1-3-14(18)19/h4-7H,1-3,8-11H2,(H,18,19) |
| SMILES | OC(=O)CCCc1ccc(cc1)N(CCCl)CCCl |
| IUPAC Name | 4-[4-[bis(2-chloroethyl)amino]phenyl]butanoic acid |
| Show More |
Product Title
Haben Sie Verbesserungsvorschläge?