Learn More
Chloramphenicol 1-O-Beta-D-Glucuronide Triethylammonium Salt, TRC
Specifica
Specifica
| Materiale o nome chimico | Chloramphenicol 1-O-beta-D-Glucuronide |
| CAS | 1013074-93-5 |
| Formula molecolare | C17H20Cl2N2O11 |
| Quantità | 1 mg |
| Sinonimo | (1R,2R)-2-[(2,2-Dichloroacetyl)amino]-3-hydroxy-1-(4-nitrophenyl)propyl β-D-Glucopyranosiduronic Acid Triethylammonium Salt |
| InChI Formula | InChI=1S/C17H20Cl2N2O11/c18-14(19)15(26)20-8(5-22)12(6-1-3-7(4-2-6)21(29)30)31-17-11(25)9(23)10(24)13(32-17)16(27)28/h1-4,8-14,17,22-25H,5H2,(H,20,26)(H,27,28)/t8-,9+,10+,11-,12-,13+,17-/m1/s1 |
| SMILES | OC[C@@H](NC(=O)C(Cl)Cl)[C@H](O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)O)c2ccc(cc2)[N+](=O)[O-] |
| IUPAC Name | (2S,3S,4S,5R,6R)-6-[(1R,2R)-2-[(2,2-dichloroacetyl)amino]-3-hydroxy-1-(4-nitrophenyl)propoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Molecular Weight (g/mol) | 499.25 |