Learn More
Chloromethyl chlorosulfate, 97%, Thermo Scientific Chemicals
Catalog Number p-3735987
Click to view available options
Quantità:
1 g
5 g
25 g
Imballaggio:
Plastic bottle
Plastic bottle packed in tin can
49715-04-0
MFCD00041547
PJBIHXWYDMFGCV-UHFFFAOYSA-N
chloromethyl chlorosulfate, chloromethyl chlorosulphate, chloromethylchlorosulfate, chloromethyl chlorosulfonate, chloromethyl chloranesulfonate, unii-n615783bnf, chloromethyl sulfochloridate, chlorosulfuric acid, chloromethyl ester, chloro chlorosulfonyloxy methane, chlorosulfuric acid chloromethyl ester
2733556
chloro(chlorosulfonyloxy)methane
C(OS(=O)(=O)Cl)Cl
49715-04-0
MFCD00041547
PJBIHXWYDMFGCV-UHFFFAOYSA-N
chloromethyl chlorosulfate, chloromethyl chlorosulphate, chloromethylchlorosulfate, chloromethyl chlorosulfonate, chloromethyl chloranesulfonate, unii-n615783bnf, chloromethyl sulfochloridate, chlorosulfuric acid, chloromethyl ester, chloro chlorosulfonyloxy methane, chlorosulfuric acid chloromethyl ester
2733556
chloro(chlorosulfonyloxy)methane
C(OS(=O)(=O)Cl)Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Chloromethyl chlorosulfate
Colore Colorless to Yellow
Spettro a infrarossi Authentic
Intervallo percentuale saggio 97%
Imballaggio Plastic bottle
Formula molecolare CH2Cl2O3S
Indice di rifrazione 1.4460 to 1.4480 (20°C, 589nm)
Quantità 1 g
Beilstein 01, III, 2593
Indice di Merck 15, 2147
Molecular Weight (g/mol) 164.984
Peso formulazione 164.99
Percent Purity ≥96.0%
Forma fisica Liquid