Learn More
Chloroparaffin C10-C13 55,5% Cl 1000 μg/mL in Cyclohexane, Dr. Ehrenstorfer
Specifications
Specifications
| Quantità | 10 mL |
| Certificazione/Conformità | ISO/IEC 17025 |
| Solvente o matrice | Cyclohexane |