Learn More
Chloroparaffin C23 41.4% Cl 100 μg/mL in Cyclohexane, Dr. Ehrenstorfer
Codice prodotto. 30315523
Cambia vista
Click to view available options
Quantità:
1 mL
Dimensione della confezione:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30315523 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30315523 Fornitore Dr. Ehrenstorfer N. del fornitore DREA11457635CY100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifica
Quantità 1 mL
Concentrazione 100 μg/mL
Certificazione/Conformità ISO/IEC 17025
Solvente o matrice Cyclohexane