missing translation for 'onlineSavingsMsg'
Learn More

Chloroparaffin C24 56% Cl 1000 μg/mL in Cyclohexane, Dr. Ehrenstorfer

Codice prodotto. 30312622
Click to view available options
Quantità:
1 mL
Dimensione della confezione:
1mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 30312622

Marca: Dr. Ehrenstorfer DREA11457630CY1000

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Quantità 1 mL
Concentrazione 1000 μg/mL
Certificazione/Conformità ISO/IEC 17025
Solvente o matrice Cyclohexane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.