missing translation for 'onlineSavingsMsg'
Learn More

Chromalite™ M Methacrylic Hydrophilic Macroporous Resins, 75 to 125 μm, Purolite™

Chromalite™ M resins are hydrophilic, highly porous with high mechanical stability, based on a rigid methacrylic backbone. Optimal for protein purification and separation.

Marca:  Chromalite LS04422-192

Dettagli aggiuntivi : Peso : 0.08000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17059483

  • 171.00€ / 100mL

Spedizione stimata: 28-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Hydrophilic, macroporous resins based on a methacrylic backbone.

  • Ion exchange chromatography
  • Typical porosity of 1000Å suitable for protein purification (up to 250 kDa)
  • Suitable for large-scale applications
  • Excellent pressure-flow properties,designed for high performance, stability and reliability
  • Purification of peptides obtained by fermentation
  • High mechanical stability
  • Uniform particle size
  • Stable to CIP (up to 1M NaOH)
  • Negligible swelling
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free
Specifica

Specifica

Sulfopropyl Methacrylate Macroporous Strong Cation Exchanger
1000 Å
Storage a 2°C to 20°C
Wet
75 to 125 μm
100 mL
Chromalite™ MS/M
Sulfopropyl / Na+
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali