Learn More
Purolite™ Chromalite™ PCG Macroporous Hydrophobic Polymeric Adsorbent
Chromalite™ PCG resins are hydrophobic porous polydivinylbenzene adsorbent. Porosity and matrix are specific for purification by reversed-phase chromatography, solid phase extraction and adsorption of biomolecules including proteins, peptides, oligonucleotides and antibiotics.
Marca:  Purolite™ LS06725-192
Dettagli aggiuntivi : Peso : 0.25000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17099553
  • 547.00€ / 250g
Spedizione stimata: 06-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Reverse phase chromatography
  • Adsorption of biomolecules
  • Solid Phase Extraction
  • Highly hydrophobic
  • High chemical stability
  • More robust than silica materials
  • Easy column packing
  • Efficient regeneration
  • Compliant with FDA regulation
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free
Specifica
Specifica
PCG1200M
2°C to 20°C
50 to 100 μm
300 to 500 Å
250 g
Wet
White to off-white spherical beads
Macroporous Polydivinylbenzene Adsorbent
1 to 14 pH
>600 m2/g
Use within 5 years from date of manufacture
65 to 80%