Learn More
Purolite™ Chromalite™ PCG Macroporous Hydrophobic Polymeric Adsorbent
Chromalite™ PCG resins are hydrophobic porous polydivinylbenzene adsorbent. Porosity and matrix are specific for purification by reversed-phase chromatography, solid phase extraction and adsorption of biomolecules including proteins, peptides, oligonucleotides and antibiotics.
Marca: Purolite™ LS04750-186
Dettagli aggiuntivi : Peso : 0.05000kg
Descrizione
- Reverse phase chromatography
- Adsorption of biomolecules
- Solid Phase Extraction
- Highly hydrophobic
- High chemical stability
- More robust than silica materials
- Easy column packing
- Efficient regeneration
- Compliant with FDA regulation
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifica
White to off-white spherical beads | |
2°C to 20°C | |
50 to 100 μm | |
75 to 150 Å | |
50 g | |
Wet |
PCG600M | |
Macroporous Polydivinylbenzene Adsorbent | |
1 to 14 pH | |
>600 m2/g | |
Use within 5 years from date of manufacture | |
55 to 70% |