Learn More
Chromate Solution, Chem Lab™
Product Code. 11909042
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11909042 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11909042 Supplier Chem-Lab Supplier No. CL01.0351.0100
  • ONLINE EXCLUSIVE PRICE
    73.00€
    35.40€ / 100mL
    Risparmia 37.60€
    51% Sconto
 Restricted Item
Spedizione stimata: 08-01-2027
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.