missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek GFP-Booster ATTO488

Codice prodotto. 17313343
Cambia vista
Click to view available options
Quantità:
10 μL
100 μL
Dimensione della confezione:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
17313343 100 μL 100µL
17303343 10 μL 10µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17313343 Fornitore Proteintech N. del fornitore gba488100
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Edge
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

GFP VHH conjugated to ATTO 488.

  • anti-GFP VHH/ Nanobody conjugated to a fluorescent dye for IF/ microscopy

Specifica

Clonality Monoclonal
Quantità 100 μL
Da utilizzare con (applicazione) Immunofluorescence (IF), Immunohistochemistry (IHC), Immunocytochemistry (ICC)
Coniugato ATTO488
Target Specificity AcGFP, Citrine, CFP, eGFP, eYFP, GFP S65T, pHluorin, sfGFP, mClover (Clover A206K), TagGFP, tagGFP2, Venus, wtGFP, YFP
Content And Storage Shipped at ambient temperature. Upon receipt store at +4™C. Stable for 6 month. Do not freeze. Protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.