missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek GST-Trap Agarose

Codice prodotto. 17363503
Change view
Click to view available options
Sufficiente per:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reazioni
100 reazioni
20 reazioni
200 reazioni
400 reazioni
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficiente per unitSize
17363503 20 Reactions 20 reazioni
17343503 10 Reactions 10 reazioni
17353503 100 Reactions 100 reazioni
17373503 200 Reactions 200 reazioni
17383503 400 Reactions 400 reazioni
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17363503 Supplier Proteintech Supplier No. sta20

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

GST-Trap is an affinity resin for IP of GST fusion proteins.

  • Consists of a GST Nanobody/ VHH coupled to agarose bead

Specifications

Content And Storage Upon receipt store at 4°C. Do not freeze.
Da utilizzare con (applicazione) IP, CoIP, ChIP, RIP
Bersaglio Glutathione S-Transferase (GST) from Schistosoma japonicum
Label or Dye Agarose
Specie Alpaca
Sufficiente per 20 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.