missing translation for 'onlineSavingsMsg'
Learn More

CIN85 Mouse anti-Human, Mouse, Rat, Clone: 3C12D2, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66571-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898565

  • 325.38€ / 150µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes an adapter protein that contains three N-terminal Src homology domains, a proline rich region and a C-terminal coiled-coil domain. The encoded protein facilitates protein-protein interactions and has been implicated in numerous cellular processes including apoptosis, cytoskeletal rearrangement, cell adhesion and in the regulation of clathrin-dependent endocytosis. Alternate splicing results in multiple transcript variants.
Specifica

Specifica

CIN85
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8R550, Q925Q9, Q96B97
Sh3kbp1
CIN85 Fusion Protein Ag2777
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG1
Western Blot
3C12D2
Unconjugated
SH3KBP1
CD2 binding protein 3, CD2BP3, CIN85, GIG10, HSB 1, MIG18, SH3KBP1
Mouse
Protein G
RUO
30011, 58194, 84357
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali