missing translation for 'onlineSavingsMsg'
Learn More

cis-1,4-Dichloro-2-butene, 95%

Codice prodotto. 11463198
Cambia vista
Click to view available options
Quantità:
25 g
100 g
500 g
Dimensione della confezione:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11463198 500 g 500g
11443198 25 g 25g
11453198 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11463198 Fornitore Thermo Scientific Alfa Aesar N. del fornitore A18956.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 1476-11-5 | C4H6Cl2 | 124.992 g/mol

It is employed as an intermediate for the production of other chemicals such as in the production of chloroprene. It is also used as a starting material in the production of adiponitrile, butane-1,4-diol, and tetrahydrofuran. It is also used in the preparation of functionalized 3,5-disubstituted cyclopent-2-enones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate for the production of other chemicals such as in the production of chloroprene. It is also used as a starting material in the production of adiponitrile, butane-1,4-diol, and tetrahydrofuran. It is also used in the preparation of functionalized 3,5-disubstituted cyclopent-2-enones.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, ether, acetone and benzene.

Notes
Moisture & Light Sensitive. Store away from oxidizing agents, heat, water/moisture and light. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1476-11-5
Formula molecolare C4H6Cl2
Numero MDL MFCD00062950
InChI Key FQDIANVAWVHZIR-UPHRSURJSA-N
Sinonimo cis-1,4-dichloro-2-butene, z-1,4-dichlorobut-2-ene, 1,4-dichloro-cis-2-butene, 2-butene, 1,4-dichloro-, z, cis-1,4-dichlorobut-2-ene, unii-bqs2752i90, ccris 2651, cis 1,4-dichlorobutene, 2-butene, 1,4-dichloro-, cis, cis-2,3-dichlorobut-2-ene
PubChem CID 6432115
IUPAC Name (Z)-1,4-dichlorobut-2-ene

Specifica

Materiale o nome chimico cis-1,4-Dichloro-2-butene
Punto di fusione -48°C
Densità 1.188
Punti di ebollizione 151°C to 153°C
Punto d'infiammabilità 55°C
Indice di rifrazione 1.49
Quantità 500 g
Numero UN UN3390
Beilstein 1719692
Sensibilità Light and moisture sensitive
Informazioni di solubilità Not miscible or difficult to mix aq. soln.. Soluble in alcohol,ether,acetone and benzene.
SMILES ClC\C=C/CCl
Molecular Weight (g/mol) 124.99
Peso formulazione 125
Percent Purity 95%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.