Learn More
CKAP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551373
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: SRLQHVEDGV LSMQVASARQ TESLESLLSK SQEHEQRLAA LQGRLEGLGS SEADQDGLAS TVRSLGETQL VLYGDVEELK RSVGELPSTV ESLQKVQEQV HTLLSQDQAQ Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 68%.
CKAP4 Polyclonal antibody specifically detects CKAP4 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifica
CKAP4 | |
Polyclonal | |
Unconjugated | |
CKAP4 | |
5630400A09Rik; 63 kDa membrane protein; 63-kDa cytoskeleton-linking membrane protein; CKAP4; Climp-63; cytoskeleton associated protein 4; cytoskeleton-associated protein 4; ERGIC-63; LPC-1; p63; transmembrane protein (63kD), endoplasmic reticulum/Golgi intermediate compartment; type-II transmembrane protein p63 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10970 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q07065 | |
CKAP4 | |
Recombinant protein corresponding to Human CKAP4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only