Learn More
Angelantoni Industrie™ Gemini Class 100 Vertical Laminar Flow Hood
Codice prodotto. 11999948
Encompass
Cambia vista
Click to view available options
Tensione:
230 V
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tensione Dimensione della confezione
11999948 230 V Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11999948 Fornitore Angelantoni Industrie™ N. del fornitore ST002190000
 Articolo soggetto a restrizioni
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Ideal vertical laminar flow hood for laboratories working in microbiology and molecular biology. Angelantoni Industrie™ Gemini Class 100 Vertical Laminar Flow Hood features a working tray and back panel of stainless steel.
  • Suitable for a variety of laboratory and manufacturing tasks.
    • Microbiology
    • Molecular biology
    • Quality control in pharmaceutical and food industries
    • Micro mechanical assembly
    • DNA/RNA amplification and thermo-cycling
  • Back panel and working tray in perforated stainless steel AISI 304L 2B
  • Front window and side walls in toughened glass
  • Lighting on working surface via 15W fluorescent lamp
  • Construction structure in epoxy-polyester powder-coated steel
    • Resistant to most common industrial disinfectants
  • Ventilation system by means of n°1 motor blower dedicated to the one-way airflow in the working chamber
  • Centrifugal type motor blower
  • Direct-driven motor with double aspiration and protection factor IP22
  • Air filtration by means of HEPA H14 type filter (refer to DIN 24184 and BS 3928) with efficiency of 99,999% at 0,3 micron particles size with DOP/DOS test (as defined to EN1822)
  • Air ventilation control by means of manual electronic regulation of motorblower revolution velocity
Specifica
Dimensioni (L x P x A) 800 x 750 x 800 mm
Altezza (sistema metrico) interno 490 mm
Tensione 230 V
Peso (sistema metrico) 70 kg
Larghezza (sistema metrico) 750 mm
Frequenza 50 Hz